3-Chloro-5-(trifluoromethyl)picolinonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-5-(trifluoromethyl)pyridine-2-carbonitrile

A160702-100G A160702-10G A160702-1G A160702-25G A160702-5G
103805-406EA 15 USD
103805-406 103843-642 77259-608 77259-610 77319-588 77319-590 77319-592
3-Chloro-5-(trifluoromethyl)picolinonitrile 97%
3-Chloro-5-(trifluoromethyl)picolinonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₃N₂
MW: 206.5524 g/mol
Boiling Pt: 250.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00664066
CAS Number: 80194-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR