3-Chloro-5-(trifluoromethyl)benzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-α,α,α-trifluoro-m-toluic acid

A169070-1G A169070-25G A169070-5G A169070-100G A169070-250MG
103806-346EA 17 USD
103806-346 103844-602 77081-670 77081-672 77081-674 77273-636 77273-638
3-Chloro-5-(trifluoromethyl)benzoic acid 98%
3-Chloro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
Flash Pt: 112.6 °C
Storage Temperature: Ambient
MDL Number: MFCD04115846
CAS Number: 53985-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR