3-Chloro-4-nitrophenol 97%
Supplier: Ambeed
|
Danger
|
A122045-100G
A122045-10G
A122045-250MG
A122045-25G
A122045-1G
A122045-5G
103800-172EA
12
USD
103800-172
76870-266
76870-268
76870-270
76870-272
77312-298
77312-300
3-Chloro-4-nitrophenol 97%
3-Chloro-4-nitrophenol
Precaución: for Research Use Only
Formula:
C₆H₄ClNO₃ Boiling Pt: 330.5 °C (760 mm Hg) Melting Pt: 120…122 °C Flash Pt: 153.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00466090 CAS Number: 491-11-2 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...