3-Chloro-4-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A117139-100G A117139-10G A117139-25G A117139-1G A117139-5G
103798-836EA 9 USD
103798-836 103824-712 76971-276 76971-278 76971-280 77297-514 77311-498
3-Chloro-4-nitrobenzoic acid 95%
3-Chloro-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
Boiling Pt: 382.7 °C (760 mm Hg)
Melting Pt: 178…180 °C
Flash Pt: 185.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01632172
CAS Number: 39608-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR