3-Chloro-4-nitrobenzenesulphonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-4-nitrobenzene-1-sulphonyl chloride

A232812-100MG A232812-250MG A232812-5G
103810-854EA 143 USD
103810-854 103850-608 77152-936 77152-938 77303-946
3-Chloro-4-nitrobenzenesulphonyl chloride 95%
3-Chloro-4-nitrobenzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₄S
Boiling Pt: 387.5 °C (760 mm Hg)
Melting Pt: 36…39 °C
Flash Pt: 188.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04973767
CAS Number: 64835-30-9
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR