3-Chloro-p-toluic acid 97%

Supplier: Ambeed
Warning

A219410-25G A219410-5G A219410-100G A219410-10G A219410-1G A219410-500G
103849-670EA 6 USD
103849-670 77088-226 77088-228 77237-614 77237-616 77237-618 77237-620
3-Chloro-p-toluic acid 97%
3-Chloro-p-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 304.5 °C (760 mm Hg)
Melting Pt: 208…209 °C
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00045841
CAS Number: 5162-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR