3-Chloro-4-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A143881-100G A143881-10G A143881-1G A143881-25G A143881-5G
103803-338EA 14 USD
103803-338 77235-126 77235-128 77316-644 77316-646 77316-648
3-Chloro-4-methoxybenzoic acid 98%
3-Chloro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
Boiling Pt: 304.8 °C (760 mm Hg)
Melting Pt: 216…218 °C
Flash Pt: 138.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00016512
CAS Number: 37908-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR