3-Chloro-4-isopropoxybenzoic acid 97%

Supplier: Ambeed
Warning

A184273-10G A184273-250MG A184273-1G A184273-25G
103815-734EA 47 USD
103815-734 103846-296 76872-852 76872-854 77301-894 77301-896
3-Chloro-4-isopropoxybenzoic acid 97%
3-Chloro-4-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C10H11ClO3
MW: 214.65 g/mol
Boiling Pt: 322.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08703674
CAS Number: 213598-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR