3-Chloro-4-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A179159-1G A179159-25G A179159-100G A179159-10G A179159-250MG
103807-402EA 6 USD
103807-402 103845-748 77083-202 77083-204 77096-776 77096-778 77096-780
3-Chloro-4-hydroxybenzonitrile 98%
3-Chloro-4-hydroxybenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClNO
Boiling Pt: 265.9 °C (760 mm Hg)
Melting Pt: 150 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD01567246
CAS Number: 2315-81-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR