3-Chloro-4-hydroxybenzoic acid 95%

Supplier: Ambeed
Warning

A120459-100G A120459-10G A120459-25G A120459-1G A120459-5G
103799-940EA 6 USD
103799-940 77136-482 77136-484 77136-486 77297-766 77312-044
3-Chloro-4-hydroxybenzoic acid 95%
3-Chloro-4-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅ClO₃
MW: 172.57 g/mol
Melting Pt: 171…173 °C
Flash Pt: 153.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002549
CAS Number: 3964-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR