3-Chloro-4-hydroxybenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A444532-25G
A444532-100G
A444532-10G
A444532-500G
A444532-5G
103827-712EA
8
USD
103827-712
103838-780
76842-562
77143-432
77143-434
77143-436
77307-326
3-Chloro-4-hydroxybenzaldehyde 97%
3-Chloro-4-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₅ClO₂ MW: 156.57 g/mol Boiling Pt: 251.1 °C (760 mm Hg) Melting Pt: 145…147 °C Flash Pt: 105.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016981 CAS Number: 2420-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...