3-Chloro-4-formylbenzonitrile 97%

Supplier: Ambeed
Warning

A212469-10G A212469-250MG A212469-1G A212469-5G
103818-206EA 42 USD
103818-206 103848-902 77012-988 77012-990 77087-366 77087-368
3-Chloro-4-formylbenzonitrile 97%
3-Chloro-4-formylbenzonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClNO
MW: 165.576 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036376
CAS Number: 58588-64-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR