(3-Chloro-4-fluorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-fluorophenylacetic acid

A440694-250MG A440694-1G A440694-5G
103827-664EA 11 USD
103827-664 103838-730 76774-038 76842-492 76842-494
(3-Chloro-4-fluorophenyl)acetic acid 95%
(3-Chloro-4-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
Boiling Pt: 302 °C (760 mm Hg)
Melting Pt: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD01631555
CAS Number: 705-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR