3-Chloro-4-fluorophenol 98%

Supplier: Ambeed
Warning

A231606-25G A231606-5G A231606-100G A231606-10G A231606-500G
103850-534EA 10 USD
103850-534 77089-422 77089-424 77288-398 77288-400 77288-402
3-Chloro-4-fluorophenol 98%
3-Chloro-4-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 225 °C (760 mm Hg)
Melting Pt: 38…40 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00002257
CAS Number: 2613-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR