3-Chloro-4-fluorobenzotrifluoride 98%

Supplier: Ambeed
Danger

Synonyms: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene

A472854-100G A472854-10G A472854-25G
103839-122EA 8 USD
103839-122 76774-298 76774-300 76843-036
3-Chloro-4-fluorobenzotrifluoride 98%
3-Chloro-4-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Boiling Pt: 136.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00042207
CAS Number: 78068-85-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR