3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A104154-100MG A104154-250MG
103796-660EA 112 USD
103796-660 103822-374 77270-370 77270-372
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline 98%
3-Chloro-4-fluoro-5-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄ClF₄N
MW: 213.56 g/mol
Boiling Pt: 235.7±40.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09033155
CAS Number: 914225-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR