3-Chloroisonicotinonitrile 98%

Supplier: Ambeed
Danger

A600629-100MG A600629-250MG A600629-1G A600629-5G A600629-10G
103828-976EA 12 USD
103828-976 103840-112 76819-394 76819-396 76844-270 76844-272 77594-828
3-Chloroisonicotinonitrile 98%
3-Chloroisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
Boiling Pt: 233.3 °C (760 mm Hg)
Flash Pt: 94.899 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663706
CAS Number: 68325-15-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR