3-Chloro-4-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-chlorobenzotrifluoride, 3-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-2-chloro-α,α,α-trifluortoluene, 4-Amino-2-chloro-α,α,α-trifluorotoluene

A362180-1G A362180-5G A362180-100G A362180-10G A362180-25G
103826-304EA 6 USD
103826-304 103837-318 76840-532 76840-534 77142-326 77142-328 77306-730
3-Chloro-4-(trifluoromethyl)aniline 97%
3-Chloro-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
Boiling Pt: 242.8 °C (760 mm Hg)
Flash Pt: 100.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278479
CAS Number: 445-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR