3-Chloro-4-(2-pyridylmethoxy)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(pyridin-2-ylmethoxy)aniline

A119481-100G A119481-10G A119481-250MG A119481-25G A119481-5G A119481-1G
103799-774EA 6 USD
103799-774 77026-288 77026-290 77026-292 77026-294 77297-690 77311-892
3-Chloro-4-(2-pyridylmethoxy)aniline 95%
3-Chloro-4-(2-pyridylmethoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClN₂O
MW: 234.68 g/mol
Boiling Pt: 402.5 °C (760 mm Hg)
Flash Pt: 197.2 °C
Storage Temperature: Ambient
MDL Number: MFCD08699389
CAS Number: 524955-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR