3-Chloro-2-nitrotoluene 98%

Supplier: Ambeed
Warning

A184125-100G A184125-10G A184125-1G A184125-5G
103815-714EA 6 USD
103815-714 103846-274 76769-644 76769-646 77083-888 77083-890
3-Chloro-2-nitrotoluene 98%
3-Chloro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
Boiling Pt: 443.8 °C (760 mm Hg)
Flash Pt: 222.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04117977
CAS Number: 5367-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR