3-Chloro-2-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
A298709-10G
A298709-250MG
A298709-1G
A298709-25G
103814-192EA
39
USD
103814-192
103835-338
76816-710
76816-712
77305-750
77305-752
3-Chloro-2-methylpyridine 97%
3-Chloro-2-methylpyridine
Precaución: for Research Use Only
Formula:
C6H6ClN Boiling Pt: 156.5 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD04114121 CAS Number: 72093-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...