3-Chloro-o-anisidine 97%

Supplier: Ambeed
Warning

A101485-100G A101485-10G A101485-25G A101485-1G A101485-5G
103796-000EA 5 USD
103796-000 103821-684 76869-154 76869-156 76869-158 77297-128 77297-130
3-Chloro-o-anisidine 97%
3-Chloro-o-anisidine

Precaución: for Research Use Only

Formula: C₇H₈ClNO
Boiling Pt: 246.0±0.0 °C (760 mm Hg)
Flash Pt: 108.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00060607
CAS Number: 51114-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR