3-Chloro-2-iodopyridine 98%

Supplier: Ambeed
Warning

A215425-1G A215425-25G A215425-5G A215425-10G A215425-250MG
103818-484EA 18 USD
103818-484 103849-190 77087-714 77087-716 77087-718 77275-302 77275-304
3-Chloro-2-iodopyridine 98%
3-Chloro-2-iodopyridine

Precaución: for Research Use Only

Formula: C5H3ClIN
MW: 239.4400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09258810
CAS Number: 77332-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR