3-Chloro-2-iodobenzoic acid 98%

Supplier: Ambeed
Warning

A121638-100MG A121638-250MG A121638-1G
103800-100EA 22 USD
103800-100 76810-504 76810-506 77312-224
3-Chloro-2-iodobenzoic acid 98%
3-Chloro-2-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
MW: 282.463 g/mol
Boiling Pt: 360.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06808824
CAS Number: 123278-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR