3-Chloro-2-iodoaniline 98%

Supplier: Ambeed
Warning

A137020-100G A137020-100MG A137020-10G A137020-250MG A137020-1G A137020-25G A137020-5G
103802-390EA 23 USD
103802-390 76984-032 76984-034 76984-036 76984-038 77315-322 77315-324 77315-326
3-Chloro-2-iodoaniline 98%
3-Chloro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₅ClIN
MW: 253.468 g/mol
Boiling Pt: 303.5 °C (760 mm Hg)
Flash Pt: 137.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738966
CAS Number: 70237-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR