3-Chloro-2-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-pyridinol

A132436-100G A132436-10G A132436-250MG A132436-1G A132436-25G A132436-5G
103801-738EA 9 USD
103801-738 76996-452 76996-454 76996-456 77314-314 77314-316 77314-318
3-Chloro-2-hydroxypyridine 98%
3-Chloro-2-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₄ClNO
MW: 129.55 g/mol
Boiling Pt: 319.5 °C (760 mm Hg)
Melting Pt: 180…182 °C
Flash Pt: 147 °C
Storage Temperature: Ambient
MDL Number: MFCD01646109
CAS Number: 13466-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR