3-Chlorosalicylic acid 98%
Supplier: Ambeed
|
Warning
|
A226676-1G
A226676-25G
A226676-5G
A226676-100G
A226676-10G
A226676-250MG
103819-260EA
10
USD
103819-260
103850-164
77088-902
77088-904
77088-906
77275-636
77275-638
77275-640
3-Chlorosalicylic acid 98%
3-Chlorosalicylic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClO₃ MW: 172.57 g/mol Boiling Pt: 291.7 °C (760 mm Hg) Melting Pt: 184…186 °C Flash Pt: 130.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010390 CAS Number: 1829-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...