3-Chlorosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-hydroxybenzaldehyde, 2-Hydroxy-3-chlorobenzaldehyde

A107494-100G A107494-10G A107494-250MG A107494-25G A107494-1G A107494-5G
103797-198EA 6 USD
103797-198 103822-966 77135-772 77135-774 77135-776 77135-778 77309-820 77309-822
3-Chlorosalicylaldehyde 98%
3-Chlorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 204.1 °C (760 mm Hg)
Melting Pt: 42…44 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973581
CAS Number: 1927-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR