3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

A873265-1G A873265-5G A873265-100G A873265-10G A873265-25G
103841-292EA 26 USD
103841-292 76845-552 76845-554 77036-756 77036-758 77036-760
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine 98%
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD00153095
CAS Number: 76041-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR