3-Chloro-2-fluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A224846-25G A224846-100G A224846-10G A224846-500G A224846-1G A224846-5G
103819-136EA 6 USD
103819-136 103850-028 77088-694 77098-036 77098-038 77098-040 77303-718 77303-720
3-Chloro-2-fluorophenylboronic acid 97%
3-Chloro-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
Boiling Pt: 315.1 °C (760 mm Hg)
Melting Pt: 223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664224
CAS Number: 352535-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR