3-Chloro-2-fluorophenol 97%

Supplier: Ambeed
Warning

A162864-100G A162864-1G A162864-25G
103843-922EA 11 USD
103843-922 77273-406 77273-408 77300-614
3-Chloro-2-fluorophenol 97%
3-Chloro-2-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFO
Boiling Pt: 225 °C (760 mm Hg)
Melting Pt: 34…38 °C
Flash Pt: 108.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01631572
CAS Number: 2613-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR