1-Chloro-2-fluoro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-2-fluoronitrobenzene

A320656-1000G A320656-25G A320656-100G A320656-10G A320656-500G A320656-5G
103810-106EA 171 USD
103810-106 76839-056 76839-058 77154-510 77154-512 77154-514 77154-516
1-Chloro-2-fluoro-3-nitrobenzene 98%
1-Chloro-2-fluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Flash Pt: 99.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00069417
CAS Number: 2106-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR