3-Chloro-2-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A133023-100G A133023-10G A133023-500G A133023-5G A133023-25G
76767-308EA 80 USD
76767-308 76767-310 76767-312 77298-600 77314-412
3-Chloro-2-fluorobenzaldehyde 98%
3-Chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClFO
Boiling Pt: 214 °C (760 mm Hg)
Flash Pt: 85.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631571
CAS Number: 85070-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR