3-Chloropyridine-2-carbonitrile 97%

Supplier: Ambeed
Danger

A414086-5G A414086-1G A414086-10G
103827-270EA 9 USD
103827-270 103838-314 76841-880 77307-170 77684-163
3-Chloropyridine-2-carbonitrile 97%
3-Chloropyridine-2-carbonitrile

Precaución: For research use only.

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
Boiling Pt: 260.7 °C (760 mm Hg)
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD03788834
CAS Number: 38180-46-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR