3-Chloro-2,5-difluoropyridine 98%

Supplier: Ambeed
Danger

A137987-1G A137987-5G A137987-250MG
103802-542EA 13 USD
103802-542 77315-542 77315-544 77670-976
3-Chloro-2,5-difluoropyridine 98%
3-Chloro-2,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MW: 149.53 g/mol
Boiling Pt: 143.3 °C (760 mm Hg)
Melting Pt: 10…12 °C
Flash Pt: 40.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09998887
CAS Number: 851179-00-5
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR