3-Chloro-2,4-difluorobenzoic acid 98%

Supplier: Ambeed
Warning

A101830-100G A101830-10G A101830-25G A101830-1G A101830-5G
103796-162EA 5 USD
103796-162 103821-854 77107-220 77107-222 77107-224 77308-748 77308-750
3-Chloro-2,4-difluorobenzoic acid 98%
3-Chloro-2,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Boiling Pt: 276.7 °C (760 mm Hg)
Melting Pt: 174…176 °C
Flash Pt: 121.1 °C
Storage Temperature: Ambient
MDL Number: MFCD01631387
CAS Number: 154257-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR