3-Chloro-2,2-bis(chloromethyl)propanoic acid 98%

Supplier: Ambeed
Warning

A320035-1G A320035-5G A320035-10G A320035-250MG
103825-224EA 21 USD
103825-224 103836-156 76839-018 76839-020 77277-694 77277-696
3-Chloro-2,2-bis(chloromethyl)propanoic acid 98%
3-Chloro-2,2-bis(chloromethyl)propanoic acid

Precaución: for Research Use Only

Formula: C₅H₇Cl₃O₂
MW: 205.4700 g/mol
Boiling Pt: 262.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00021685
CAS Number: 17831-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR