3-Chloro-1H-pyrazol-4-amine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A325428-250MG A325428-25G A325428-5G
103825-404EA 15 USD
103825-404 76989-194 77683-969 77683-970
3-Chloro-1H-pyrazol-4-amine 97%, Ambeed.Inc
3-Chloro-1H-pyrazol-4-amine

Precaución: For research use only.

Formula: C₃H₄ClN₃
MW: 117.54 g/mol
Boiling Pt: 344.5±22.0 °C (760 mm Hg)
Melting Pt: 149…150 °C
Flash Pt: 162.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23701740
CAS Number: 103286-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR