3-Carboxy-N,N,N-trimethylpropan-1-aminium chloride 97%

Supplier: Ambeed
Warning

A410057-1G A410057-25G A410057-5G A410057-10G A410057-250MG
103827-208EA 31 USD
103827-208 103838-254 76841-768 76841-770 76841-772 77143-030 77143-032
3-Carboxy-N,N,N-trimethylpropan-1-aminium chloride 97%
3-Carboxy-N,N,N-trimethylpropan-1-aminium chloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00011905
CAS Number: 6249-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR