3-Butyne-1-ol 98%

Supplier: Ambeed
Danger

Synonyms: 4-Hydroxy-1-butyne, But-3-yn-1-ol, 3-Butyn-1-ol, 1-Hydroxy-3-butyne

A426437-25G A426437-10G A426437-5G
103838-522EA 5 USD
103838-522 76842-178 76877-210 77307-238
3-Butyne-1-ol 98%
3-Butyne-1-ol

Precaución: for Research Use Only

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 128.8
Melting Pt: -64
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00002955
CAS Number: 927-74-2
UN: 1987
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR