1-Butyl-3-methylimidazoliumtetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: 3-Butyl-1-methyl-1H-imidazol-3-ium tetrafluoroborate, 1-n-Butyl-3-methylimidazolium tetrafluoroborate, [BMIM] [BF4]

A141019-10G A141019-5G
103808-614EA 306 USD
103808-614 77027-516 77299-180
1-Butyl-3-methylimidazoliumtetrafluoroborate 98%
1-Butyl-3-methylimidazoliumtetrafluoroborate

Precaución: for Research Use Only

Formula: C₈H₁₅BF₄N₂
MW: 226.02 g/mol
Melting Pt: 71 °C
Flash Pt: 288 °C
Storage Temperature: Ambient
MDL Number: MFCD03095449
CAS Number: 174501-65-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR