1-Butyl-3-methylimidazolium bromide 97%

Supplier: Ambeed
Warning

A108648-100G A108648-25G A108648-500G A108648-5G A108648-10G
103808-000EA 334 USD
103808-000 103823-158 76780-972 76780-974 76780-976 77283-764 77570-500
1-Butyl-3-methylimidazolium bromide 97%
1-Butyl-3-methylimidazolium bromide

Precaución: for Research Use Only

Formula: C₈H₁₅BrN₂
Melting Pt: 65…75 °C
Storage Temperature: Ambient
MDL Number: MFCD03427611
CAS Number: 85100-77-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR