3-Bromothioanisole 95%

Supplier: Ambeed
Warning

A582742-25G A582742-5G A582742-100G A582742-10G A582742-1G
103840-020EA 12 USD
103840-020 76844-150 76844-152 77144-746 77144-748 77144-750
3-Bromothioanisole 95%
3-Bromothioanisole

Precaución: for Research Use Only

Formula: C₇H₇BrS
Boiling Pt: 278.106 °C (760 mm Hg)
Flash Pt: 121.995 °C
Storage Temperature: Ambient
MDL Number: MFCD00041395
CAS Number: 33733-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR