3-Bromothieno[3,2-c]pyridin-4-amine 97%

Supplier: Ambeed
Warning

A143417-250MG A143417-1G
103803-278EA 197 USD
103803-278 76811-864 77316-542
3-Bromothieno[3,2-c]pyridin-4-amine 97%
3-Bromothieno[3,2-c]pyridin-4-amine

Precaución: for Research Use Only

Formula: C7H5BrN2S
MW: 229.0970 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08275115
CAS Number: 799293-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR