3-Bromothieno[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

A212091-100MG A212091-250MG A212091-1G A212091-5G
103818-164EA 52 USD
103818-164 103848-862 76814-582 76814-584 77303-216 77303-218
3-Bromothieno[2,3-b]pyridine 97%
3-Bromothieno[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C7H4BrNS
MW: 214.0824 g/mol
Boiling Pt: 299.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11869739
CAS Number: 28988-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR