6-Amino-3-bromoquinoline 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromoquinolin-6-amine

A132000-100MG A132000-250MG A132000-1G
103801-676EA 63 USD
103801-676 76823-044 76823-046 77314-218
6-Amino-3-bromoquinoline 98%
6-Amino-3-bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Boiling Pt: 359 °C (760 mm Hg)
Melting Pt: 154…155 °C
Flash Pt: 170.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07757603
CAS Number: 7101-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR