3-Bromoquinolin-5-amine 95%

Supplier: Ambeed
Warning

A401522-1G A401522-250MG
103827-056EA 60 USD
103827-056 103838-092 76841-554 77168-300 77168-302
3-Bromoquinolin-5-amine 95%
3-Bromoquinolin-5-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD20327979
CAS Number: 116632-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR