3-Bromopicolinic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid
A115820-100G
A115820-10G
A115820-25G
A115820-500G
A115820-1G
103798-630EA
5
USD
103798-630
103824-490
77284-492
77284-494
77284-496
77284-498
77297-400
3-Bromopicolinic acid 97%
3-Bromopicolinic acid
Precaución: for Research Use Only
Formula:
C₆H₄BrNO₂ Boiling Pt: 315.7 °C (760 mm Hg) Melting Pt: 141…144 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01320380 CAS Number: 30683-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...