3-Bromopyridazine 97%

Supplier: Ambeed
Warning

A300699-25G A300699-250MG A300699-10G
103814-278EA 40 USD
103814-278 103835-428 76838-114 77032-488 77601-358
3-Bromopyridazine 97%
3-Bromopyridazine

Precaución: for Research Use Only

Formula: C₄H₃BrN₂
MW: 158.9840 g/mol
Boiling Pt: 270 °C (760 mm Hg)
Flash Pt: 117.075 °C
Storage Temperature: Freezer
MDL Number: MFCD10688587
CAS Number: 88491-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR