3-Bromopyrazolo[1,5-a]pyrimidine 97%
Supplier: Ambeed
|
Warning
|
A138968-10G
A138968-250MG
A138968-1G
A138968-5G
103802-678EA
12
USD
103802-678
77272-384
77272-386
77315-740
77315-742
3-Bromopyrazolo[1,5-a]pyrimidine 97%
3-Bromopyrazolo[1,5-a]pyrimidine
Precaución: for Research Use Only
Formula:
C6H4BrN3 Melting Pt: 150…152 °C Storage Temperature: Ambient |
MDL Number:
MFCD09832893 CAS Number: 55405-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...